Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccndbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010761.2 | NP_034891.2 | 3 | 316 | ||
GenBank mRNA | AK144883.1 | - | 3 | 293 | ||
AK154285.1 | - | 3 | 270 | |||
AK160205.1 | - | 3 | 282 | |||
AK163150.1 | - | 3 | 229 | |||
AK167345.1 | - | 3 | 281 | |||
AK186314.1 | - | 2 | 93 | |||
BC001984.1 | - | 3 | 252 | 3489973 | ||
BC005802.1 | - | 2 | 325 | 3707919 | ||
CO810054.1 | - | 3 | 323 | |||
U50734.1 | - | 3 | 234 |