Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccndbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010761.2 | NP_034891.2 | 4 | 371 | ||
GenBank mRNA | AK144883.1 | - | 4 | 348 | ||
AK154285.1 | - | 4 | 325 | |||
AK160205.1 | - | 4 | 337 | |||
AK163150.1 | - | 4 | 284 | |||
AK167345.1 | - | 4 | 336 | |||
AK186314.1 | - | 3 | 148 | |||
BC001984.1 | - | 4 | 307 | 3489973 | ||
BC005802.1 | - | 3 | 380 | 3707919 | ||
CO810054.1 | - | 4 | 378 | |||
U50734.1 | - | 4 | 289 |