Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccndbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010761.2 | NP_034891.2 | 8 | 706 | ||
GenBank mRNA | AK144883.1 | - | 8 | 683 | ||
AK154285.1 | - | 8 | 660 | |||
AK160205.1 | - | 8 | 672 | |||
AK163150.1 | - | 8 | 619 | |||
AK167345.1 | - | 8 | 671 | |||
BC001984.1 | - | 8 | 642 | 3489973 | ||
BC004673.1 | - | 2 | 126 | |||
BC005802.1 | - | 7 | 715 | 3707919 | ||
U50734.1 | - | 8 | 624 |