Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Marco | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010766.2 | NP_034896.1 | 15, 16 | 1564 | ||
GenBank mRNA | AK156388.1 | - | 15, 16 | 1238 | ||
AK159094.1 | - | 2, 3 | 69 | |||
AK159967.1 | - | 15, 16 | 1246 | |||
AK169230.1 | - | 15, 16 | 1240 | |||
BC132338.1 | - | 15, 16 | 1504 | 40130615 | ||
BC137939.1 | - | 15, 16 | 1504 | 8860958 | ||
U18424.1 | - | 15, 16 | 1416 |