Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Marco | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010766.2 | NP_034896.1 | 16 | 1615 | ||
GenBank mRNA | AK156388.1 | - | 16 | 1289 | ||
AK159094.1 | - | 3 | 120 | |||
AK159967.1 | - | 16 | 1297 | |||
AK169230.1 | - | 16 | 1291 | |||
BC132338.1 | - | 16 | 1555 | 40130615 | ||
BC137939.1 | - | 16 | 1555 | 8860958 | ||
U18424.1 | - | 16 | 1467 |