Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Matk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285853.1 | NP_001272782.1 | 6 | 844 | ||
NM_001285854.1 | NP_001272783.1 | 6 | 841 | |||
NM_001285855.1 | NP_001272784.1 | 6 | 909 | |||
NM_010768.2 | NP_034898.1 | 7 | 977 | |||
GenBank mRNA | AK013606.1 | - | 6 | 658 | ||
AK018182.1 | - | 6 | 695 | |||
BC060086.1 | - | 5 | 1080 | |||
BC111864.1 | - | 6 | 655 | 40046807 | ||
D45243.1 | - | 6 | 909 | |||
L27738.1 | - | 7 | 731 | |||
S77473.1 | - | 6 | 656 | |||
U05210.1 | - | 6 | 630 |