Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Matk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285853.1 | NP_001272782.1 | 11 | 1389 | ||
NM_001285854.1 | NP_001272783.1 | 11 | 1386 | |||
NM_001285855.1 | NP_001272784.1 | 11 | 1454 | |||
NM_010768.2 | NP_034898.1 | 12 | 1522 | |||
GenBank mRNA | AK013606.1 | - | 11 | 1203 | ||
AK079816.1 | - | 8 | 1259 | |||
AK158883.1 | - | 8 | 1324 | |||
BC060086.1 | - | 7 | 2521 | |||
BC111864.1 | - | 9 | 1371 | 40046807 | ||
D45243.1 | - | 11 | 1454 | |||
L27738.1 | - | 12 | 1276 | |||
S77473.1 | - | 11 | 1201 | |||
U05210.1 | - | 11 | 1175 |