Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Matn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010769.2 | NP_034899.2 | 4 | 767 | ||
GenBank mRNA | AK014342.1 | - | 4 | 767 | ||
BC047140.1 | - | 4 | 734 | 6315509 | ||
U35035.1 | - | 4 | 728 |