Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Meis1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193271.1 | NP_001180200.1 | 4 | 1142 | ||
NM_010789.3 | NP_034919.1 | 4 | 1142 | |||
GenBank mRNA | AB221660.1 | - | 3 | 405 | ||
AK032947.1 | - | 3 | 953 | |||
AK049720.1 | - | 3 | 954 | |||
AK132298.1 | - | 4 | 922 | |||
AK134321.1 | - | 4 | 1137 | |||
AK140748.1 | - | 4 | 1146 | |||
AK168684.1 | - | 4 | 1134 | |||
BC023689.1 | - | 4 | 1138 | 5344307 | ||
U33629.1 | - | 4 | 853 | |||
U33630.1 | - | 4 | 853 |