Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Meis1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193271.1 | NP_001180200.1 | 11 | 1814 | ||
NM_010789.3 | NP_034919.1 | 11 | 1814 | |||
GenBank mRNA | AB221660.1 | - | 10 | 1077 | ||
AK134321.1 | - | 11 | 1809 | |||
AK140748.1 | - | 11 | 1818 | |||
AK146680.1 | - | 7 | 622 | |||
AK168684.1 | - | 11 | 1807 | |||
U33629.1 | - | 11 | 1525 | |||
U33630.1 | - | 11 | 1525 |