Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Meis1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193271.1 | NP_001180200.1 | 13 | 2892 | ||
NM_010789.3 | NP_034919.1 | 12 | 2797 | |||
GenBank mRNA | AK146680.1 | - | 9 | 1700 | ||
AK168684.1 | - | 13 | 2885 | |||
BC034136.1 | - | 1 | 1986 | |||
U33629.1 | - | 13 | 2610 | |||
U33630.1 | - | 12 | 2515 |