Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Melk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010790.2 | NP_034920.2 | 4 | 403 | ||
GenBank mRNA | AK010498.1 | - | 4 | 370 | ||
AK011932.1 | - | 4 | 403 | |||
AK129076.1 | - | 4 | 356 | |||
AK137000.1 | - | 4 | 428 | |||
AK145021.1 | - | 4 | 338 | |||
AK145316.1 | - | 4 | 312 | |||
AK161029.1 | - | 4 | 412 | |||
AK164138.1 | - | 4 | 370 | |||
AK167457.1 | - | 4 | 327 | |||
BC020416.1 | - | 3 | 238 | |||
BC085276.1 | - | 4 | 436 | 30443236 | ||
L76158.1 | - | 4 | 334 | |||
X95351.1 | - | 4 | 341 |