Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Melk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010790.2 | NP_034920.2 | 5 | 481 | ||
GenBank mRNA | AK010498.1 | - | 5 | 448 | ||
AK011932.1 | - | 5 | 481 | |||
AK129076.1 | - | 5 | 434 | |||
AK137000.1 | - | 5 | 506 | |||
AK145021.1 | - | 5 | 416 | |||
AK145316.1 | - | 5 | 390 | |||
AK161029.1 | - | 5 | 490 | |||
AK164138.1 | - | 5 | 448 | |||
AK167457.1 | - | 5 | 405 | |||
BC085276.1 | - | 5 | 514 | 30443236 | ||
L76158.1 | - | 5 | 412 | |||
X95351.1 | - | 5 | 419 |