Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290504.1 | NP_001277433.1 | 953 | |||
NM_001290505.1 | NP_001277434.1 | 1177 | ||||
NM_001290506.1 | NP_001277435.1 | 1063 | ||||
NM_001290512.1 | NP_001277441.1 | 397 | ||||
NM_010797.3 | NP_034927.2 | 1060 | ||||
GenBank mRNA | AF026565.1 | - | 953 | |||
AK160120.1 | - | 399 | ||||
AY540025.1 | - | 106 | ||||
AY540026.1 | - | 106 | ||||
AY540027.1 | - | 585 | ||||
AY540030.1 | - | 122 | ||||
AY540031.1 | - | 122 | ||||
AY540032.1 | - | 1737 | ||||
AY540033.1 | - | 189 | ||||
AY540035.1 | - | 808 | ||||
AY540036.1 | - | 278 | ||||
AY540037.1 | - | 122 | ||||
BC053704.1 | - | 1030 | 30058736 | |||
Y14848.1 | - | 1014 |