Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Minpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010799.2 | NP_034929.1 | 5 | 1404 | ||
GenBank mRNA | AB041574.1 | - | 5 | 1237 | ||
AF046908.1 | - | 5 | 1404 | |||
AK128911.1 | - | 5 | 1283 | |||
AK151486.1 | - | 5 | 1249 | |||
BC021437.1 | - | 5 | 1305 | 5039704 |