Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010808.3 | NP_034938.3 | 4 | 614 | ||
GenBank mRNA | AB021226.1 | - | 4 | 559 | ||
AJ010262.1 | - | 4 | 560 | |||
AY622976.1 | - | 3 | 203 |