Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010808.3 | NP_034938.3 | 8, 9 | 1645 | ||
GenBank mRNA | AB021226.1 | - | 8, 9 | 1590 | ||
AJ010262.1 | - | 8, 9 | 1591 | |||
BM950641.1 | - | 2, 3 | 388 |