Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGES | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004878.4 | NP_004869.1 | 3 | 1540 | ||
XM_011519210.2 | XP_011517512.1 | 4 | 1901 | |||
GenBank mRNA | AF010316.1 | - | 3 | 1498 | ||
AF217965.1 | - | 4 | 1619 | |||
AF217972.1 | - | 1 | 1047 | |||
AK127663.1 | - | 1 | 2682 | |||
BC008280.1 | - | 3 | 1528 | 3623516 |