Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289442.1 | NP_001276371.1 | 8 | 2257 | ||
NM_010815.3 | NP_034945.1 | 7 | 2352 | |||
GenBank mRNA | AF055465.1 | - | 7 | 2352 | ||
AK030809.1 | - | 1 | 833 | |||
AK156939.1 | - | 7 | 2331 |