Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010832.5 | NP_034962.2 | 2, 3 | 266 | ||
GenBank mRNA | AF117066.2 | - | 2, 3 | 268 | ||
AK089368.1 | - | 2, 3 | 349 | |||
AK146208.1 | - | 2, 3 | 203 | |||
AK148898.1 | - | 2, 3 | 245 | |||
BC010226.1 | - | 2, 3 | 227 | 3501126 |