Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001043355.2 | NP_001036820.2 | 3 | 1204 | ||
NM_010837.3 | NP_034967.2 | 4 | 2300 | |||
GenBank mRNA | AK018193.2 | - | 4 | 1780 | ||
AK154455.1 | - | 4 | 1176 | |||
BY259340.1 | - | 3 | 426 | |||
Y16032.1 | - | 3 | 1144 |