Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mthfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161798.1 | NP_001155270.1 | 12 | 4138 | ||
NM_010840.3 | NP_034970.2 | 12 | 4061 | |||
GenBank mRNA | AK149795.1 | - | 13 | 3054 | ||
AK150159.1 | - | 12 | 4139 | |||
BC051017.1 | - | 14 | 2730 | 6308248 | ||
BC052466.1 | - | 12 | 4105 | 6834886 | ||
BC100550.1 | - | 4 | 2529 |