Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039657.2 | NP_001034746.1 | 4 | 816 | ||
NM_001039658.2 | NP_001034747.1 | 4 | 816 | |||
NM_001286557.1 | NP_001273486.1 | 5 | 900 | |||
NM_001286558.1 | NP_001273487.1 | 4 | 831 | |||
GenBank mRNA | AB057423.1 | - | 4 | 804 | ||
AF329359.1 | - | 4 | 841 | |||
AF329360.1 | - | 5 | 910 | |||
AK015724.1 | - | 4 | 757 | |||
AK015732.1 | - | 4 | 789 | |||
AK161511.1 | - | 4 | 817 | |||
BC024377.1 | - | 4 | 792 | 4976492 | ||
U67176.1 | - | 6 | 833 | |||
U77383.1 | - | 4 | 453 |