Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mxi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008542.2 | NP_001008542.2 | 5 | 806 | ||
NM_001008543.2 | NP_001008543.1 | 5 | 493 | |||
NM_010847.3 | NP_034977.1 | 5 | 646 | |||
GenBank mRNA | AK140465.1 | - | 5 | 596 | ||
AK144219.1 | - | 5 | 688 | |||
AK154538.1 | - | 5 | 796 | |||
BC064453.1 | - | 5 | 791 | 6810025 | ||
D31824.1 | - | 5 | 646 | |||
L38821.1 | - | 5 | 499 | |||
L38822.1 | - | 5 | 518 | |||
U24673.1 | - | 5 | 479 | |||
U24674.1 | - | 5 | 428 |