Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myef2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162417.1 | NP_001155889.1 | 6 | 706 | ||
NM_001162418.1 | NP_001155890.1 | 6 | 706 | |||
NM_010852.2 | NP_034982.2 | 6 | 706 | |||
GenBank mRNA | AF483504.1 | - | 5 | 432 | ||
AF483505.1 | - | 5 | 432 | |||
AK012133.1 | - | 6 | 708 | |||
AK048404.1 | - | 6 | 673 | |||
AK129336.1 | - | 6 | 618 | |||
AK142363.1 | - | 6 | 622 | |||
AK143230.1 | - | 6 | 642 | |||
BY711324.1 | - | 6 | 704 | |||
U13262.1 | - | 2 | 147 |
Gene Symbol: Gm9833 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC060946.1 | - | 1 | 460 | 6741255 |