Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161817.2 | NP_001155289.1 | 2, 3 | 355 | ||
NM_001290982.1 | NP_001277911.1 | 2, 3 | 355 | |||
NM_010863.4 | NP_034993.2 | 2, 3 | 355 | |||
GenBank mRNA | AK053754.1 | - | 3, 4 | 763 | ||
AK138265.1 | - | 2, 3 | 290 | |||
AK141207.1 | - | 2, 3 | 330 | |||
AK149409.1 | - | 2, 3 | 302 | |||
AK161062.1 | - | 2, 3 | 319 | |||
BC026826.1 | - | 2, 3 | 303 | |||
BC046300.1 | - | 2, 3 | 301 | 6487332 | ||
BC054786.1 | - | 2, 3 | 305 | 6821232 | ||
L00923.1 | - | 2, 3 | 324 |