Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myom1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083934.1 | NP_001077403.1 | 12, 13 | 1901 | ||
NM_010867.2 | NP_034997.2 | 12, 13 | 1901 | |||
GenBank mRNA | AJ012072.1 | - | 12, 13 | 1934 | ||
AK052539.1 | - | 12, 13 | 1868 | |||
BC060378.1 | - | 11, 12 | 1806 | 4980452 | ||
Z22866.1 | - | 12, 13 | 1880 |