Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010887.2 | NP_035017.2 | 4, 5 | 469 | ||
GenBank mRNA | AF124785.1 | - | 4, 5 | 419 | ||
AK011124.1 | - | 4, 5 | 457 | |||
AK214655.1 | - | 1, 2 | 81 | |||
BC004618.1 | - | 4, 5 | 427 | 3585543 |