Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENTPD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128930.2 | NP_001122402.1 | 10, 11 | 1630 | ||
NM_004901.4 | NP_004892.1 | 10, 11 | 1654 | |||
GenBank mRNA | AJ131358.1 | - | 10, 11 | 1551 | ||
AK300464.1 | - | 10, 11 | 1562 | |||
BC034477.2 | - | 10, 11 | 1594 | 4824559 | ||
DB091432.1 | - | 2, 3 | 374 |