Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfatc2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010900.3 | NP_035030.2 | 5 | 791 | ||
GenBank mRNA | AK005947.1 | - | 5 | 782 | ||
AK030107.1 | - | 5 | 793 | |||
BC113761.1 | - | 5 | 792 | 40054445 | ||
U76759.1 | - | 5 | 781 |