Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010926.5 | NP_035056.1 | 5 | 784 | ||
GenBank mRNA | AF052621.1 | - | 5 | 786 | ||
AK042704.1 | - | 5 | 652 | |||
AK076710.1 | - | 7 | 1653 | |||
AK143131.1 | - | 5 | 745 | |||
AK148190.1 | - | 5 | 1295 | |||
AK161438.1 | - | 5 | 760 | |||
AK164130.1 | - | 5 | 785 | |||
BC009103.1 | - | 5 | 740 | 3482942 |