Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nov | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010930.4 | NP_035060.1 | 5 | 1953 | ||
GenBank mRNA | AK039481.1 | - | 6 | 1874 | ||
AK081944.1 | - | 6 | 2117 | |||
AK155048.1 | - | 5 | 1972 | |||
BC003774.1 | - | 6 | 1889 | 3494896 | ||
X96585.1 | - | 5 | 1952 | |||
Y09257.1 | - | 5 | 1941 |