Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS9D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004913.2 | NP_004904.2 | 2 | 277 | ||
XM_005256329.4 | XP_005256386.1 | 2 | 337 | |||
XM_005256330.4 | XP_005256387.1 | 2 | 154 | |||
XM_006721350.2 | XP_006721413.1 | 2 | 337 | |||
XM_011523476.2 | XP_011521778.1 | 2 | 337 | |||
XM_011523477.2 | XP_011521779.1 | 1 | 1569 | |||
XM_011523478.2 | XP_011521780.1 | 1 | 1569 | |||
XM_011523479.2 | XP_011521781.1 | 2 | 337 | |||
XM_011523480.2 | XP_011521782.1 | 2 | 191 | |||
GenBank mRNA | AB018551.1 | - | 1 | 318 | ||
AI458261.1 | - | 2 | 277 | |||
BC052626.1 | - | 1 | 658 |