Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr1i2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010936.3 | NP_035066.1 | 5 | 951 | ||
GenBank mRNA | AF031814.1 | - | 5 | 766 | ||
AK018630.1 | - | 5 | 951 | |||
BC120796.1 | - | 5 | 698 | 40129719 | ||
BC137780.1 | - | 5 | 698 | 8860799 |