Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Numbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010950.2 | NP_035080.2 | 8 | 1151 | ||
GenBank mRNA | AK147523.1 | - | 6 | 6938 | ||
BC059915.1 | - | 7 | 1740 | |||
BC068116.1 | - | 8 | 978 | 6809443 | ||
BC098097.1 | - | 8 | 992 | 6403012 | ||
U96441.1 | - | 9 | 1032 |