Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oit3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010959.2 | NP_035089.1 | 9 | 2127 | ||
GenBank mRNA | AF356506.1 | - | 9 | 2074 | ||
AF517106.1 | - | 8 | 1946 | |||
AF517107.1 | - | 8 | 1946 | |||
AK040570.1 | - | 9 | 2128 | |||
AK050081.1 | - | 9 | 2094 | |||
AK153440.1 | - | 5 | 1369 | |||
AY180915.1 | - | 9 | 2051 |