Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P4ha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011030.2 | NP_035160.1 | 3 | 327 | ||
GenBank mRNA | AK030574.1 | - | 3 | 259 | ||
AK045008.1 | - | 3 | 256 | |||
AK149127.1 | - | 3 | 219 | |||
AK160798.1 | - | 3 | 256 | |||
AK161212.1 | - | 3 | 260 | |||
AK169726.1 | - | 3 | 260 | |||
BC009654.1 | - | 3 | 250 | 3484815 | ||
BE985798.1 | - | 3 | 327 | |||
BY721363.1 | - | 3 | 308 | |||
U16162.1 | - | 2 | 97 |