Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P4ha2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011031.2 | NP_035161.2 | 13, 14 | 1635 | ||
GenBank mRNA | CD563665.1 | - | 6, 7 | 493 | ||
U16163.1 | - | 12, 13 | 1522 |