Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pax8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011040.4 | NP_035170.1 | 12 | 1849 | ||
GenBank mRNA | AK050717.1 | - | 12 | 1825 | ||
AK052610.1 | - | 12 | 1826 | |||
BC020526.1 | - | 12 | 1816 | 4239835 | ||
X57487.1 | - | 12 | 1806 |