Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103165.1 | NP_001096635.1 | 5 | 389 | ||
NM_001103166.1 | NP_001096636.1 | 5 | 389 | |||
NM_001174073.1 | NP_001167544.1 | 5 | 365 | |||
NM_011042.2 | NP_035172.2 | 5 | 389 | |||
GenBank mRNA | AK012297.1 | - | 5 | 237 | ||
AK161527.1 | - | 5 | 213 | |||
AK169268.1 | - | 5 | 483 | |||
BC020478.1 | - | 4 | 134 | |||
BQ900329.1 | - | 5 | 365 | |||
EU604548.1 | - | 4 | 144 | |||
L19661.1 | - | 5 | 276 |