Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011049.4 | NP_035179.1 | 16 | 1790 | ||
GenBank mRNA | AK051425.1 | - | 16 | 1791 | ||
AK141083.1 | - | 10 | 810 | |||
AK143518.1 | - | 16 | 1818 | |||
AK145483.1 | - | 16 | 1820 | |||
AK166188.1 | - | 16 | 1999 | |||
BC011069.1 | - | 16 | 1533 | 4006745 | ||
BC013663.1 | - | 16 | 1849 | 4208974 | ||
X69025.1 | - | 16 | 1812 |