Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011049.4 | NP_035179.1 | 16 | 2218 | ||
GenBank mRNA | AK051425.1 | - | 16 | 2219 | ||
AK141083.1 | - | 10 | 1238 | |||
AK145483.1 | - | 16 | 2248 | |||
AK209172.1 | - | 1 | 364 | |||
AK210955.1 | - | 1 | 364 | |||
AK219069.1 | - | 1 | 364 | |||
BC011069.1 | - | 16 | 1976 | 4006745 | ||
X69025.1 | - | 16 | 2240 |