Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Padi3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011060.4 | NP_035190.3 | 10 | 1180 | ||
GenBank mRNA | AB013849.1 | - | 10 | 1181 | ||
AK220522.1 | - | 9 | 1422 |