Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Per1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159367.1 | NP_001152839.1 | 5 | 797 | ||
NM_011065.4 | NP_035195.2 | 5 | 812 | |||
GenBank mRNA | AB002108.1 | - | 4 | 626 | ||
AF022992.1 | - | 5 | 851 | |||
AK081813.1 | - | 5 | 810 | |||
AK148202.1 | - | 5 | 804 | |||
AK154900.1 | - | 5 | 793 | |||
AK172958.1 | - | 5 | 797 | |||
AK182563.1 | - | 2 | 155 | |||
BC039768.1 | - | 4 | 757 | 4163009 | ||
BC091645.1 | - | 5 | 799 | 30355259 |