Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Per3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289877.1 | NP_001276806.1 | 6, 7 | 1219 | ||
NM_001289878.1 | NP_001276807.1 | 7, 8 | 1268 | |||
NM_011067.3 | NP_035197.2 | 6, 7 | 1219 | |||
GenBank mRNA | AB013605.1 | - | 6, 7 | 1026 | ||
AF050182.1 | - | 6, 7 | 1143 | |||
AJ007377.1 | - | 6, 7 | 1026 | |||
AK036345.1 | - | 6, 7 | 1136 | |||
AK166514.1 | - | 6, 7 | 1220 | |||
BC141032.1 | - | 6, 7 | 817 | 9055644 | ||
BC145178.1 | - | 7, 8 | 866 |