Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Per3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289877.1 | NP_001276806.1 | 10 | 1609 | ||
NM_001289878.1 | NP_001276807.1 | 11 | 1658 | |||
NM_011067.3 | NP_035197.2 | 10 | 1609 | |||
GenBank mRNA | AB013605.1 | - | 11 | 1416 | ||
AF050182.1 | - | 10 | 1533 | |||
AJ007377.1 | - | 11 | 1416 | |||
AK036345.1 | - | 10 | 1526 | |||
AK045115.1 | - | 1 | 1008 | |||
AK084610.1 | - | 3 | 2577 | |||
AK166514.1 | - | 11 | 1843 | |||
AK173261.1 | - | 1 | 937 | |||
BC141032.1 | - | 11 | 1207 | 9055644 | ||
BC145178.1 | - | 12 | 1256 |