Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Per3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289877.1 | NP_001276806.1 | 17 | 3178 | ||
NM_001289878.1 | NP_001276807.1 | 18 | 3227 | |||
NM_011067.3 | NP_035197.2 | 17 | 3175 | |||
GenBank mRNA | AB013605.1 | - | 18 | 2982 | ||
AF050182.1 | - | 17 | 3099 | |||
AJ007377.1 | - | 18 | 2982 | |||
AK084610.1 | - | 9 | 4024 | |||
AK139878.1 | - | 3 | 976 | |||
AK166514.1 | - | 18 | 3406 | |||
AK173261.1 | - | 5 | 4465 | |||
BC141032.1 | - | 18 | 2776 | 9055644 | ||
BC145178.1 | - | 19 | 2825 |