Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfdn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011070.3 | NP_035200.2 | 3 | 261 | ||
GenBank mRNA | BC026839.1 | - | 3 | 248 | 4219635 | |
BC049606.1 | - | 3 | 261 | 6535749 | ||
Y17393.1 | - | 3 | 251 |