Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011072.4 | NP_035202.1 | 1, 2 | 294 | ||
GenBank mRNA | AK011066.1 | - | 1, 2 | 279 | ||
AK028117.1 | - | 1, 2 | 245 | |||
AK150780.1 | - | 1, 2 | 251 | |||
AK152491.1 | - | 1, 2 | 280 | |||
AK160191.1 | - | 1, 2 | 280 | |||
AV448567.1 | - | 1, 2 | 295 | |||
BC002080.1 | - | 1, 2 | 267 | 3490976 | ||
X14425.1 | - | 1, 2 | 183 |