Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011076.2 | NP_035206.2 | 16 | 2108 | ||
GenBank mRNA | M24417.1 | - | 10 | 1457 | ||
M30697.1 | - | 16 | 2122 | |||
M33581.1 | - | 16 | 2107 |